Parts Washing Baskets, with Locking Lid, Marlin Steel Wire Products
Fournisseur: Marlin Steel Wire Products
Certificats
A propos de cet article
Stainless steel mesh baskets with lid that locks.
- Electropolished finish
- With 30 mesh openings per linear inch
Great for material handling and parts washing/ultrasonic cleaning.
Customers Who Bought This Also Bought