VWR® Contour™ Storage Cabinets with Glass Doors
Fournisseur: VWR International
Certificats
A propos de cet article
VWR Contour® tall units, wall-mounted units, or wall-mounted corner units.
- Cold-rolled steel
- Glazed glass doors
Wall-mounted units are available with sliding or swinging glass doors.
Certifications: Meets SEFA recommended practices.
Ordering information: Accessories not included.
Caution: Two person moving only.
Customers Who Bought This Also Bought