Ward's® Weigh Bucket
# de catalogue 470314-478
Fournisseur: MTC BIO, INC.
Spécifications
- Description:Ward's® weigh bucket
- Cat. no.:470314-478
Spécifications
A propos de cet article
The weigh boat for liquids.
- Easy-grip handle
- 40 ml capacity
- Convenient pour spout
- Stackable
Ths polyethylene weigh bucket is a new type of weighing vessel that safely transports, contains and pours liquids, powders and small solids. The handle and spout make them easy to use, practically eliminating spillage and making controlled-rate pouring possible.
Caution: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Customers Who Bought This Also Bought