A propos de cet article
The VWR® A-series Balances with NTEP Certificate and Calibration Certificate provide legal-for-trade certification along with full traceability for regulatory and compliance needs.
- Internal calibration
- 2×RS-232, USB (A and B), and Wi-Fi communication interfaces
- Five year warranty
- Tool-free assembly and diassembly of the 3-door draft shield on analytical models
The VWR® A-series balances with NTEP certification and Calibration certificate are Class I and Class II certified balances featuring an NTEP Certificate of Conformance indicating the devices meet the requirements of Handbook 44. These balances also include a certificate of calibration showing the full testing data from an ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory using NIST-traceable weights. Featuring internal calibration, a multitude of communication options, and built in data protections, the VWR® A-series balances provide advanced features for any application. The large, back-lit LCD display allows for easy viewing of the weighing results. Data safety is provided via the internal memory allowing for recording up to 100000 weighings. The analytical models feature a spacious 3-door draft shield that allows for tool-free assembly and disassembly of the weighing chamber. The VWR® A-series balances with NTEP certification and Certificate of Calibration are the answer for organizations demanding precision, performance and full traceability in legal-for-trade applications.
The 0.01 and 0.1 mg readability models feature a fixed 3-door draft shield (which can not be removed). The 0.001 g readability models feature a removable, glass draft shield.
Spécifications
- Catalog No:
- 76458-534
- 76378-394
- 76458-536
- 76458-538
- 76458-544
- 76458-546
- 76458-540
- 76458-542
- 76458-532
- 76378-396
- 76378-398
- Operating temperature range:
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- Plug type:
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Communication interface:
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- Voltage:
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V