Savant™ SpeedVac™ Modular Vacuum Concentrator, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Thermo Scientific™ Savant™ SpeedVac™ SPD300DDA vacuum concentrator is a large-capacity vacuum concentrator with a modular design.
- Kit
- Large Capacity
- Variety of rotors
- Heated glass safety cover
- PTFE coated aluminum chamber
The SpeedVac™ SPD300DDA is ideal for drying aggressive organic solvents, strong acids, bases, and combinatorial chemistry solvents. It is capable of drying larger quantities of samples at one time and can support high-volume sample preparation needs.
Ordering information: Supplied as different packages, please see description for details of what is included in specific kits/packages.
Specifications
- Catalog No:
- 76311-114
- 76210-584
- 76210-580
- W×D×H:
- 483×406.4×457 mm (19×16×18")
- 483×406.4×457 mm (19×16×18")
- 483×406.4×457 mm (19×16×18")