KIMBLE® HI-VAC® Low Hold-Up Valves, with Extended Tip, without Plug, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Valves with straight low hold up extended tip.
- Sidearms are bent to minimize holdup of liquids
- O-rings made of viton
PTFE shaft seals protect the viton O-rings from contact with the system. Seal design reduces outgassing.
Customers Who Bought This Also Bought