Savant™ SpeedVac™ Modular Vacuum Concentrator, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
The Thermo Scientific™ Savant™ SpeedVac™ SPD140DDA vacuum concentrator is a medium-capacity vacuum concentrator with a modular design.
- Kit
- Medium capacity
- Variety of rotors
- Heated glass safety cover
- PTFE coated aluminum chamber
The SpeedVac™ SPD140DDA is ideal for drying aggressive organic solvents, strong acids, bases, and combinatorial chemistry solvents.
Ordering information: Supplied as different packages, please see description for details of what is included in specific kits/packages.
Spécifications
- Catalog No:
- 76210-578
- 76210-574
- W×D×H:
- 330×457×356 mm (13×18×14")
- 330×457×356 mm (13×18×14")