Savant™ SpeedVac™ Modular Vacuum Concentrators, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
The Thermo Scientific™ Savant™ SpeedVac™ SPD210 vacuum concentrator is a large-capacity vacuum concentrator with a modular design, ideal for drying aqueous and organic solvents.
- Kit
- Medium capacity
- Variety of rotors
- Glass safety cover
- PTFE coated aluminum chamber
The SpeedVac™ SPD210 is capable of drying larger quantities of samples at one time and will support your high-volume sample preparation needs.
Ordering information: Supplied as different packages, please see description for details of what is included in specific kits/packages.
Spécifications
- Catalog No:
- 76210-400
- 76210-568
- 76210-564
- W×D×H:
- 483×660×457 mm (19×26×18")