Savant™ SpeedVac™ Intergrated Vacuum Concentrator, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
The Savant SpeedVac SPD1030 system is designed for medium capacity (120×1.5 ml tubes and other formats) and the SpeedVac SPD2030 system is ideal for higher capacity (200×1.5 ml tubes and other formats).
- Fully integrated
- Medium capacity
- Variety of rotors
- Radiant glass cover
- PTFE coated aluminum chamber
A variety of rotors are available to support sample formats from micro-centrifuge tubes, vials and microplates to large-volume flasks. The unit will fit in a standard laboratory fume hood.
Spécifications
- Catalog No:
- 76210-548
- W×D×H:
- 635×660×406 mm (25×26×16")