Avantor® ACE® Guard Column Cartridge Holders
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Stainless steel Avantor® ACE® stand-alone and integral guard cartridge holders are used for connecting guard cartridges to the LC column to provide column protection.
Integral holders connect directly to the analytical column. Stand-alone holders require the use of a suitable connector.
Customers Who Bought This Also Bought