Avantor® ACE® Phenyl Ultra-Inert, HPLC Columns, 3 µm
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
These ultra inert Avantor® ACE® Phenyl-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
- Designed for peptide and protein separations
- Excellent peak shape even at extremely low levels of additives such as TFA, making them ideal for LC/MS
- Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Spécifications
- Catalog No:
- 76390-068
- 76390-010
- 76382-594
- 76390-012
- 76390-108
- 76390-112
- 76390-046
- 76390-052
- 76390-136
- Carbon load:
- 5.30%
- 5.30%
- 5.30%
- 5.30%
- 5.30%
- 5.30%
- 5.30%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 400 bar (5801 psi)
- 400 bar (5802 psi)
- 400 bar (5801 psi)
- 400 bar (5801 psi)
- 400 bar (5801 psi)
- 400 bar (5801 psi)
- 400 bar (5801 psi)
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- USP code:
- L11
- L11
- L11
- L11
- L11
- L11
- L11