KIMBLE® ULTRA-WARE® Standard Fittings Kit, DWK Life Sciences
# de catalogue KT953882-1000
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:Standard fittings kit
- Cat. no.:KT953882-1000
- Supplier No.:953882-1000
Spécifications
A propos de cet article
This kit contains all of the tube fittings, adapters and solvent inlet filters required to plumb any ULTRA-WARE® cap to virtually all HPLC pumps.
Adapter, F 1/4-28 to F Luer (420407-2003), adapter, M 1/4-28 to M Luer (420407-2113), union, 5/16-24 to 1/4-28 (420818-3305), tubing nut, 1/4-28, for ¹/₈" OD (420821-0018), tubing nut, M6, for ¹/₈" OD (420821-0618), tubing nut, 5/16-24, for ³/₁₆" OD (420821-3519), ferrule, for ¹/₈" OD (420822-0018), ferrule, for ³/₁₆" OD (420822-1019), ferrule, for 1.8 mm OD (420822-2007), inlet filter, for ¹/₈" OD tubing (953916-0000), inlet filter, BOTB, for ¹/₈" OD tubing (953916-1002), adapter, 1/4-28 to 1/4" hose barb (953919-0014). Not recommended for use with THF or chlorinated hydrocarbons.
Customers Who Bought This Also Bought