KIMBLE® Fittings Kit, DWK Life Sciences
# de catalogue KT953882-0000
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:Start-up fittings kit
- Cat. no.:KT953882-0000
- Supplier No.:953882-0000
Spécifications
A propos de cet article
Kit consists of 20 of ¹/₈" O.D FEP PTFE tubing, 20 CTFE ferrules and 20 (¹/₄"-28) ETFE nuts.
Customers Who Bought This Also Bought