ContactUsLinkComponent
KIMBLE® Fittings Kit, DWK Life Sciences
KIMBLE® Fittings Kit, DWK Life Sciences
# de catalogue KT953882-0000
HPLC Cap System Economy 3-Hole ULTRA-WARE
KIMBLE® Fittings Kit, DWK Life Sciences
# de catalogue KT953882-0000
Spécifications
  • Description:
    Start-up fittings kit
  • Cat. no.:
    KT953882-0000
  • Supplier No.:
    953882-0000
Spécifications
A propos de cet article
Kit consists of 20 of ¹/₈" O.D FEP PTFE tubing, 20 CTFE ferrules and 20 (¹/₄"-28) ETFE nuts.
Customers Who Bought This Also Bought