ContactUsLinkComponent
Adapter, Vacuum, DWK Life Sciences
Adapter, Vacuum, DWK Life Sciences
# de catalogue KT747140-0000
Labsets kits Organic Chemistry Microscale Threaded Research MICROFLEX
Adapter, Vacuum, DWK Life Sciences
# de catalogue KT747140-0000
Spécifications
  • Description:
    Vacuum adapter
  • GPI Finish:
    13-425, 20-400
  • Cat. no.:
    KT747140-0000
  • Supplier No.:
    747140-0000
Spécifications
A propos de cet article
Glassware is manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
  • Ideal for use to 200 °C
Customers Who Bought This Also Bought