Adapter, Vacuum, DWK Life Sciences
# de catalogue KT747140-0000
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:Vacuum adapter
- GPI Finish:13-425, 20-400
- Cat. no.:KT747140-0000
- Supplier No.:747140-0000
Spécifications
A propos de cet article
Glassware is manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
- Ideal for use to 200 °C
Customers Who Bought This Also Bought