KIMBLE® Bidwell-Sterling Distillation Receivers, Moisture Test, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Bidwell-Sterling designed for the determination of moisture in foods and organics.
- Made to ASTM specifications E123
- Also conforms to many methods of the American oil chemists society
Manufactured from 33 expansion, low extractable.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought