Electrode Cleaning and Storage Solutions, Oakton®, Cole Parmer
Supplier: Argos Technologies
Certificates
About this item
Clean electrodes make clean measurements.
- Easily remove grease, protein, and other deposits with cleaning solution
- Make electrode last longer—store in a storage solution
Cleaning solution contains pepsin. After cleaning, keep your electrodes in proper working condition with our storage solution.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.