WHEATON® High Performance Crimping Tool, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Crimping tool is designed with inter-changeable jaws, so that you only need one tool.
- Fastest and most powerful WHEATON crimping tool
- Strong enough for all steel and magentic caps
- Designed with external power source and cord (no battery)
- Uses interchangeable jaw sets
- Stores up to 9 programs for different caps and seals
- Optional base and mounting kit
The jaw sets are available as crimper and decappers which fit standard lab vials seals with aluminum or two-part aluminum/steel caps. This crimping tool has the ability to store separate programs for different caps and cap sizes. The tools are ergonomically designed to reduce strain and arm injury associated with many manual crimping tools
Not for use with Flip-up caps.
Customers Who Bought This Also Bought