DURAN® Silicone Sealing Disc, PTFE Coated, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
High leak tightness is achieved through the use of our PTFE coated silicone seal.
- Autoclavable
- PTFE-coated platinum-cured silicone
Customers Who Bought This Also Bought