DURAN® Silicone Seal, PTFE Coated (VMQ), DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These seals are suitable for PBT screw caps and tamper-evident screw caps.
- Autoclavable
- Peroxidically-cured silicon
- Heat resistance 130 °C (vapor) and 200 °C (dry heat)
Good chemical resistance due to PTFE coating.
Customers Who Bought This Also Bought