DURAN® YOUTILITY, Bottle Tags, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
These bottle tags are used for easy customization and unambiguous bottle identification.
- Fits thread GL 45
- Securely attaches around the neck of the bottle
- Elastic for flexibility
- Wide range of colors available
- Autoclavable at 121 °C
The colorful silicone GL 45 bottle tags can be easily and securely attached around the neck of the YOUTILITY bottle. The Bottle Tags are available in several distinct colors for easy customization and unambiguous bottle identification. The elastic bottle tags will fit around any DIN GL 45 bottle neck, and can even be applied with the screw cap in place.
Customers Who Bought This Also Bought