KIMBLE® KONTES® HI-VAC® Valved NMR Tubes, DWK Lifesciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
These tubes are designed to improve sample handling during the investigation of volatile, air-sensitive, or toxic compounds.
- KONTES® HI-VAC® valve maximizes sample recovery
- Greaseless valve features a borosilicate glass body and PTFE plug so that the sample is always in contact with inert materials
- Rotationally symmetrical design, with two lateral ports located near the valve seat, permits use at higher temperatures
- FKM O-ring seal is used to connect the valved NMR tube to a vacuum system
Suitable for use in instruments with field strengths greater than 360 MHz.
Customers Who Bought This Also Bought