KIMBLE® Inlet Gas Adapters with Septum, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Inlet gas adapters have a standard taper joint.
- Manufactured from 33 expansion, low extractable borosilicate glass
Sidearm has a 2 mm bore PTFE stopcock with a port for gas introduction and a rubber-stoppered port on the top tube for syringe retrieval.
Delivery: Supplied with KT821001-0002 PTFE stopcock and KT882311-0000 septum stopper.
Customers Who Bought This Also Bought