KIMBLE® Westergren Sedimentation Tube, DWK Life Sciences
# de catalogue 70720-035
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Length:240 mm
- Description:Westergren sedimentation tube, blue
- Cat. no.:70720-035
- Inner Diameter:2.55 mm
- Supplier No.:67G70720-035
Spécifications
A propos de cet article
ESR tubes measure the rate at which red blood cells settle out of plasma. Westergren sedimentation rate tubes may be used for the standard procedure, by drawing blood in a liquid sodium citrate vacuum tube, or the modified procedure, using a saline-diluted EDTA vacuum tube.
- Precision-drawn glass and accurate calibrations ensure that the results are accurate with the standard normal values
- Manufactured from 90 expansion soda lime glass conforming to USP Type III requirements
Customers Who Bought This Also Bought