KIMAX® KimCote® Heavy-Duty Solution Bottles, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
KimCote® plastic safety coated solution bottles with carboy clamp and silicone stopper assemblies for solution bottles.
- Neck is tooled for a #12 stopper
- Five gallon size is designed from Federal Specification DD-B-597
Customers Who Bought This Also Bought