KIMBLE® HI-VAC® Straight Valves, with CTFE Plug and Tip O-rings, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
These straight high-vacuum valves are used for vacuum applications up to 5×10-7 Torr.
- Designed with a precision external thread for fine control
- Can be used as a bleed up to ½ turn
- Full open position is at 1½ turns
- Primary seal is made with a FKM O-ring on a tapered seat
- Non-rotating CTFE shaft provides for improved outgassing at system temperatures up to 200 °C
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought