Claisen Adapter, Threaded, DWK Life Sciences
# de catalogue KT747110-0000
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:Claisen Adapter, 13-425
- Height:50 mm
- Cat. no.:KT747110-0000
- Supplier No.:747110-0000
Spécifications
A propos de cet article
This adapter converts single-neck flasks or vials to three-neck vessels for chemical reactions such as Grignard reactions.
- Allows easy, vacuum-tight connections of syringes for liquid addition, thermometers or gas inlet tubes
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought