KIMBLE® Schlenk tubes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These Schlenk tubes are designed to minimize the loss of volatile solvents while manipulating under normal conditions.
- Working capacity is 5 ml to less than 1 ml solvent or 100 mg to less than 1 mg substrate
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Customers Who Bought This Also Bought