Vacuum Manifolds, Inert Gas, Double, with PTFE Valves, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Double vacuum gas manifold is for connecting inert gas and vacuum sources.
- Medium wall tubing: ³/₄"
- Rob braces gives additional strength
- Opened upper left and lower right beams
- Manufactured from 33 expansion, low extractable borosilicate glass
100 mm spaced ports with serrated hose connections.
PTFE vacuum valves (4 mm) at 45° angle is for grease free operation and to allow easier manipulation.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought