DURAN® High Temperature PBT Screw Caps, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
DURAN® red PBT closed screw caps with PTFE protected seal protect important materials in the lab.
- High leak tightness through use of PTFE coated silicone seal
- More chemically resistant than PP screw cap
- Autoclavable at 121 °C
Permitting clean, drip-free use.
Ordering information: Matching ETFE pouring rings can be ordered.
Customers Who Bought This Also Bought