Vacuum Manifolds, Inert Gas, with PTFE Valves, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These manifolds are used to connect inert gas and vacuum sources to sidearm stopcocks (three-place, four-place or five-place manifolds). Which is manufactured from 33 expansion, low extractable borosilicate glass.
- Distance between valves is 100 mm
- Ports have 9 mm OD openings
- Sample contacts only PTFE and borosilicate glass
Each stopcock has a metering groove cut into the plug to provide fine control of vacuum sources and inert gases. Beams are ¹/₂" (12.7 mm) OD, with upper-left and lower-right ends open.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought