Drying Tube, Bent, DWK Life Sciences
# de catalogue KT746095-0000
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- ST Joint No.:14/10
- Width:85 mm
- Cat. no.:KT746095-0000
- Overall Length:45 mm
- Bulb O.D.:30 mm
- Supplier No.:746095-0000
Spécifications
A propos de cet article
This drying tube is used with an unsealed system to protect moisture-sensitive reactions from the atmosphere.
- Modified drying tube with an expansion bulb for CaCl₂
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought