Vacuum Manifolds, Inert Gas, Double, with Vacuum Chambers, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
General purpose double bank manifold for use with vacuum and inert gas.
- Medium wall tubing size: ¹/₂"
- Manufactured from 33 expansion, low extractable borosilicate glass
- Stopcocks are 4 mm bore glass with lower vacuum chamber and spaced 85 mm
Top right connection is for inert gas and lower left connection is for vacuum.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought