KIMBLE® Thermometer Inlet Adapters, DWK Lifesciences
# de catalogue 89002-178
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:Rubber adapters, 6/case
- Cat. no.:89002-178
- Hole Size:¹/₄"
- Supplier No.:44870R-99
Spécifications
A propos de cet article
These adapters are suitabel for use with plain stem thermometers and gas inlet tubes.
Customers Who Bought This Also Bought