A propos de cet article
Permanent autoclave-resistant labels withstand various sterilization procedures, including in steam autoclaves, dry heat ovens, gamma irradiators, and ethylene oxide gas sterilizers.
- Temperature range: –60 to +150 °C (–76 to +302 °F)
- Ideal for use in repeated steam autoclave sterilization cycles, gamma irradiation (up to 50 KGy), and ethylene oxide sterilization
- Versatile labels adhere to a variety of surfaces including metal, plastic, and glass
- Printable with all major thermal transfer printer brands
- Waterproof and weatherproof
- RR-class resin ribbon recommended
High temperature-resistant permanent labels for identifying various sterilization containers and surfaces that will undergo autoclaving and dry heat sterilization. Temperature ranges from –60 to +150 °C (–76 to +302 °F). The labels can be applied to metal, plastic, and glass surfaces, and will withstand up to 20 autoclave sterilization cycles at 121 °C (250 °F) and 16.5 psi pressure.
The labels and the printout can withstand high temperature exposure, as well as gamma irradiation (up to 50 KGy) and ethylene oxide sterilization. Ideal for identifying containers and surfaces that need to be sterilized inside autoclaves, dry heat sterilizing equipment, and with gamma irradiators. Waterproof, weatherproof, thermal transfer printable.