KIMBLE® NMR Sample Tubes, 3 mm, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Designed for high resolution work with NMR spectrometers.
- High-quality tubes are ideal for sealing directly to vacuum manifolds, joints, or valves
- Tubes feature a sandblasted marking spot
Manufactured from ASTM Type 1 borosilicate glass under rigid quality control to meet exacting specifications for wall chambers and variation.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Tubes for 800 and 900MHz NMR are also available, contact your Avantor sales representative for more information.
Disposable and thrift-grade tubes do not have caps attached, high-quality tubes are supplied with caps attached.
Disposable and thrift-grade tubes do not have caps attached, high-quality tubes are supplied with caps attached.
Specifications
- Catalog No:
- KT897820-0008
- KT897830-0000
- KT897800-0000
- KT897835-0000
- KT897840-0000
- KT897805-0000
- KT897840-0008
- Ext.Ø:
- 0.1186"
- 0.1186"
- 0.1186"
- 0.1186"
- 0.1186"
- 0.1186"
- 0.1186"
- Wall thickness:
- 0.01118"
- 0.01118"
- 0.01118"
- 0.01118"
- 0.01118"
- 0.01118"
- 0.01118"