VWR® Blood Draw Chair
Catalog # 76353-720
Supplier: VWR International
Specifications
- Width:29"
- Description:Phlebotomy chair
- Height:37"
- Length:32"
- Model:76353-720
- Frame colour/finish:White
- Seat/back color:Blue
- Seat/back material:Vinyl
- Frame material:Steel
- Seat dimensions:30" (19" internal seat)
- Weight capacity:400 lbs.
- Cat. no.:76353-720
- Weight:52 lbs.
Specifications
About this item
Space-saving blood drawing chair with padded, height adjustable arms for optimal comfort and a heavy duty steel frame for optimal strength and durability.
- Adjustable arms provide optimum vein puncture level
- Heavy duty epoxy-coated welded steel frame
- CAL-133 certified medical grade vinyl
Space-saving design with a small footprint. This chair offers a fully upholstered foam padded seat and armpads for extra comfort, height adjustable arms for optimal vein puncture, and a heavy duty steel frame for extra strength and durability, all while comfortably fitting in spaces 30" or more. A 400 lb. patient weight capacity and a 19" internal seat width offers all the flexibility you need caring for your patients while conserving precious exam room space.
Certifications: CE certified.
BodyTextAccessories: Accessories not included.
Delivery: Time to manufacture: 3 to 4 weeks, delivery time: 4 to 6 weeks.
Customers Who Bought This Also Bought