KIMBLE® HI-VAC® Right-Angle Valves, with CTFE Plug and Tip O-rings, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
This right-angle high-vacuum valve is for use in vacuum applications up to 5 x 10-⁷ torr.
- Designed with a precision external thread for fine control
- Can be used as a bleed up to ¹/₂ turn
- Full open position is at 1-¹/₂ turns
- Primary seal is made with a FKM O-ring on a tapered seat
- Non-rotating CTFE shaft provides for improved outgassing at system temperatures up to 200 °C
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought