KIMBLE® HI-VAC® Straight Valves, with Glass Plug and Tip O-rings, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
This valve uses a glass plug (as an alternative to PTFE or CTFE) for applications where the 'out-gassing' plug material is undesirable.
- Designed with a precision external thread for fine control
- This valve can be used as a bleed up to ¹/₂ turn
- Fully open position is at 1-¹/₂ turns
- Greater solvent resistance
All plugs are totally interchangeable with other HI-VAC® counterparts.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery: Supplied with FKM O-rings.
Customers Who Bought This Also Bought