Polytron® Generator, PT-DA/2MEC, Kinematica
Fournisseur: Kinematica
Certificats
A propos de cet article
M-design rotor with knives for pre-cutting. Outside blades pre-crush samples that are larger than the rotors diameter. Subsequently, they can be dispersed inside the generator. Dispersing generators are made from high-alloy stainless steel 316L, electropolished, Ra≤1.6 μm as standard.
- Dispersing of samples with larger diameters made simple
- Standard dispersing, emulsifying and suspending tasks
Customers Who Bought This Also Bought