DURAN® GLS 80® Stirred Reactors, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
The GLS 80® stirred reactor is suitable for a wide range of laboratory mixing processes.
- Autoclavable
- The stirrer unit is interchangeable
- Drive for the stirrer may be provided by a standard commercial magnetic stirrer
The connections (2×GL 14 and 2×GL 18) provided permit addition or removal of media from the bottle during the mixing process. The whole unit can be autoclaved and is therefore suitable for biological applications. By using components from the GLS 80® connection system, an additional media bottle (Tubing outer diameter: 1.6 – 12.0 mm) can be connected or a sterile pressure equalizer attached.
The variable stirrer shaft can be used in DURAN® GLS 80® laboratory glass bottles (1000 and 2000 ml) and provides notably improved mixing in comparison with standard magnetic stir bars. The stirrer unit is interchangeable and can be used up to 500 rpm is possible.
Customers Who Bought This Also Bought