DURAN® Premium Caps, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Cap is moulded from a pigment free polymer to reduce the risk of leaching.
- Wide usable temperature range from –196 to +200 °C
- Very high chemical resistance
- Colorless PFA, with PTFE faced silicone cap liner
- Autoclavable
Typical applications: Due to its exceptional properties, it is ideal for applications in the pharmaceutical industry, storage of aggressive materials, and demanding sterilisation processes such as hot air sterilisation or depyrogenation.
Customers Who Bought This Also Bought