HyFlex® 74-050 Cut-Resistant Gloves, Ansell
# de catalogue 76377-478
Fournisseur: Ansell Healthcare
Spécifications
- Ambidextrous/Hand-specific:Hand-specific
- Glove Color:White,Gray
- Glove size:9
- Length:235 - 305 mm (9.25 - 12")
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Cuff style:Knit Wrist
- Cat. no.:76377-478
Spécifications
A propos de cet article
A close, comfortable fit with the protection of a heavyweight glove.
- Excellent levels of cut protection: ANSI Level A6
- Incorporates Tuff-Cuff™ anti-shrink, extended cuff construction
- Excellent protection in food processing applications
- Good for most knife-hand and some off-hand applications
- Available in white and gray
Offering a high level of cut protection and lightweight comfort, HyFlex® 74-050 series gloves provide superior dexterity and can be used as a liner under another glove.
Certifications: ANSI 3 Abrasion; ANSI A6 Cut; EN 388:2016 4X2XF; EN 388:2003 454X; EN 420:2003 + A1:2009.
Packaging: Gloves are packaged 12 pieces per case.
Customers Who Bought This Also Bought