A propos de cet article
Specifically designed for easy marking and identification inside Dewar-type liquid nitrogen tanks (–196 °C) and laboratory freezers (–80 °C/–112 °F).
- Specifically designed to adhere to metal surfaces, such as stainless steel
- Resists extreme cryogenic temperatures (–196 °C/–321 °F, in liquid or vapor phase liquid nitrogen)
- Printable with all common thermal transfer barcode printer brands
- Versatile design allows labels to withstand temperatures up to 100° C (212 °F)
MetaliTAG™ cryogenic labels are designed for the identification of metal surfaces in liquid nitrogen Dewars, including aluminum cryogenic vial canes, metal racks for cryo and freezer boxes, as well as aluminum cassettes and metal sleeves. These specialty labels are made from a cryogenic material that has been tested and approved for applying to stainless steel and aluminum with subsequent immersion and storage inside liquid phase or vapor phase liquid nitrogen. The MetaliTAG™ cryo labels can be printed on with all common brands of thermal transfer barcode printers.
MetaliTAG™ cryogenic labels have been tested for stainless steel and aluminum with subsequent immersion and storage inside liquid or vapor phase liquid nitrogen.