KIMBLE® Vacuum Hydrolysis Tubes, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Versatile tube for use where a vacuum environment is desired.
- Useful in protein hydrolysis and amino acid separations
- Other applications include sealed tube reactions, freeze drying and concentration
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, class A requirements.
Caution: Not recommended for use over 150 °C.
Customers Who Bought This Also Bought