DURAN® Screw Caps with Aperture, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
These red PBT screw caps are suitable for silicone seal for piercing (Septa).
- Maximaum temperature: 180 °C
- Autoclavable at 121 °C
Typical applications include injection or removal of media.
Customers Who Bought This Also Bought