Properties of Materials Kit
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Fully investigate density, buoyancy and specific gravity.
- Allows students to make a wide range of investigations with only a small number of items
- Contains two sets of three metal cylinders - aluminum, copper, and steel
In one set the cylinders all have equal masses, and in the other they have equal volumes. The measuring instruments - cylinder, spring scale, and caliper - measure volume, mass, and length.
This combination of items allows for a wide range of basic investigations, addressing the concepts of mass, volume, density and specific gravity and extending to an exploration of buoyancy.
Delivery: Includes manual.
Customers Who Bought This Also Bought