AC Meters
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Full range of AC meters.
- Ideal for student labs
- Full-sized
- Fully enclosed
Our meters are mounted in black plastic cases. Meters have a scale length of 75 mm, offer ±2.5% accuracy and come with five-way binding posts and an external zero-setting screw. Meters are not protected against prolonged or significant overload. Stackable for storage.
Customers Who Bought This Also Bought