DURAN® Vacuum Desiccator, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Vacuum-tight, made from DURAN® borosilicate glass 3.3. To accelerate drying, a vacuum can be applied via the stopcock. Spare parts such as lids, bases, stopcocks and caps can be interchanged (observe DN).
- Without porcelain plate
- With NOVUS standard ground joint
- Junction tube in the lid (NS 24/29), stopcock, and flat flange
Typical applications: drying of moist samples and storage of moisture-sensitive substances.
Customers Who Bought This Also Bought