WHEATON® All Purpose Reagent Reservoirs, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These reservoirs are particularly well suited for single and multichannel pipetting. Various shape, material and capacity provides ideal solutions for many applications.
- Polystyrene
- 8 or 12 channel reservoir
- 48 ml capacity
Customers Who Bought This Also Bought