A propos de cet article
Color chemical-resistant labels designed for use with containers exposed to harsh chemical and solvents.
- Will withstand direct exposure to harsh chemicals and solvents
- Durable adhesive ensures labels will not wrinkle or peel
- Available in a variety of colors for color coding and easy identification
These solvent-resistant thermal-transfer labels are available in an assortment of colors and are resistant to xylene, acetone, toluene, as well as commercial cleaners. These labels can withstand immersion in xylene for up to 15 minutes without detaching or the print fading and smudging. Ideal for labeling glass or plastic chemical storage containers, as well as microscope slide boxes, plastic flasks, and metal or plastic chemical drums. These color labels allow for color coding and easy identification thanks to their unique coating, designed to provide optimal chemical resistance in addition to high-quality printouts of alphanumeric text, serial numbers, and 1D and/or 2D barcodes.
Permanent color labels are perfect for labeling glass and plastic containers that will be used to store harsh chemicals and solvents or will come into contact with them. Removable labels are coated with a glove-friendly adhesive that ensures theys can be removed without leaving any glue residue behind, making them ideal for labeling glass, polypropylene, and metal surfaces.